siRNA Details
| Gene Symbol: Cops7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108807.1 | NP_001102277.1 | 2 | 354 | ||
| XM_006245414.2 | XP_006245476.1 | 1 | 805 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cops7b| Gene Symbol: Cops7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108807.1 | NP_001102277.1 | 2 | 354 | ||
| XM_006245414.2 | XP_006245476.1 | 1 | 805 | |||