siRNA Details
| Gene Symbol: Naa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135839.1 | NP_001129311.1 | 7 | 542 | ||
| XM_006229591.2 | XP_006229653.1 | 7 | 852 | |||
| XM_006229592.2 | XP_006229654.1 | 7 | 651 | |||
| GenBank mRNA | FQ235221.1 | - | 7 | 486 | ||

Naa10
Genomic Map