siRNA Details
| Gene Symbol: Smg9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014243.1 | NP_001014265.1 | 11 | 1230 | ||
| XM_006228450.2 | XP_006228512.1 | 11 | 2161 | |||
| GenBank mRNA | BC087051.1 | - | 11 | 1230 | 7115791 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smg9| Gene Symbol: Smg9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014243.1 | NP_001014265.1 | 11 | 1230 | ||
| XM_006228450.2 | XP_006228512.1 | 11 | 2161 | |||
| GenBank mRNA | BC087051.1 | - | 11 | 1230 | 7115791 | |