siRNA Details
| Gene Symbol: Mamstr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001076631.5 | XP_001076631.2 | 2 | 1002 | ||
| XM_006229210.2 | XP_006229272.1 | 2 | 1387 | |||
| XM_006229211.2 | XP_006229273.1 | 2 | 946 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mamstr| Gene Symbol: Mamstr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001076631.5 | XP_001076631.2 | 2 | 1002 | ||
| XM_006229210.2 | XP_006229272.1 | 2 | 1387 | |||
| XM_006229211.2 | XP_006229273.1 | 2 | 946 | |||