siRNA Details
| Gene Symbol: Tysnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108932.1 | NP_001102402.1 | 3 | 1509 | ||
| GenBank mRNA | BC160877.1 | - | 3 | 1472 | 7373508 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tysnd1| Gene Symbol: Tysnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108932.1 | NP_001102402.1 | 3 | 1509 | ||
| GenBank mRNA | BC160877.1 | - | 3 | 1472 | 7373508 | |