siRNA Details
| Gene Symbol: Lcn8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128183.1 | NP_001121655.1 | 3 | 273 | ||
| XM_006233632.2 | XP_006233694.2 | 3 | 414 | |||
| GenBank mRNA | DQ537491.1 | - | 3 | 259 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lcn8| Gene Symbol: Lcn8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128183.1 | NP_001121655.1 | 3 | 273 | ||
| XM_006233632.2 | XP_006233694.2 | 3 | 414 | |||
| GenBank mRNA | DQ537491.1 | - | 3 | 259 | ||