siRNA Details
| Gene Symbol: Rbm38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108965.1 | NP_001102435.1 | 1 | 289 | ||
| XM_006235704.2 | XP_006235766.1 | 1 | 284 | |||
| XM_008762528.1 | XP_008760750.1 | 1 | 287 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm38| Gene Symbol: Rbm38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108965.1 | NP_001102435.1 | 1 | 289 | ||
| XM_006235704.2 | XP_006235766.1 | 1 | 284 | |||
| XM_008762528.1 | XP_008760750.1 | 1 | 287 | |||