siRNA Details
| Gene Symbol: LOC100910205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750592.3 | XP_003750640.3 | 3 | 825 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100910205| Gene Symbol: LOC100910205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750592.3 | XP_003750640.3 | 3 | 825 | ||