siRNA Details
Gene Symbol: Dst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244713.2 | XP_006244775.1 | 2, 3 | 449 | ||
XM_008766967.1 | XP_008765189.1 | 2, 3 | 444 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244713.2 | XP_006244775.1 | 2, 3 | 449 | ||
XM_008766967.1 | XP_008765189.1 | 2, 3 | 444 |