siRNA Details
| Gene Symbol: Mageh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013250.1 | NP_001013268.1 | 1 | 632 | ||
| GenBank mRNA | BC087574.1 | - | 1 | 632 | 7306512 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mageh1| Gene Symbol: Mageh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013250.1 | NP_001013268.1 | 1 | 632 | ||
| GenBank mRNA | BC087574.1 | - | 1 | 632 | 7306512 | |