siRNA Details
| Gene Symbol: Zc4h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126374.1 | NP_001119846.1 | 3, 4 | 508 | ||
| GenBank mRNA | BC162002.1 | - | 3, 4 | 508 | 9088857 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc4h2| Gene Symbol: Zc4h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126374.1 | NP_001119846.1 | 3, 4 | 508 | ||
| GenBank mRNA | BC162002.1 | - | 3, 4 | 508 | 9088857 | |