siRNA Details
| Gene Symbol: Ift81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199120.1 | NP_954551.1 | 7 | 735 | ||
| GenBank mRNA | AY345164.1 | - | 7 | 735 | ||
| FQ225832.1 | - | 7 | 765 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ift81| Gene Symbol: Ift81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199120.1 | NP_954551.1 | 7 | 735 | ||
| GenBank mRNA | AY345164.1 | - | 7 | 735 | ||
| FQ225832.1 | - | 7 | 765 | |||