siRNA Details
| Gene Symbol: Olr14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000695.1 | NP_001000695.1 | 1 | 329 | ||
| Gene Symbol: LOC100910049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748918.1 | XP_003748966.1 | 329 | |||

Olr14
Genomic Map