siRNA Details
| Gene Symbol: Btla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213630.1 | NP_998795.1 | 2 | 165 | ||
| XM_006248341.2 | XP_006248403.1 | 2 | 400 | |||
| GenBank mRNA | AY590499.1 | - | 2 | 165 | ||
| BC092588.1 | - | 2 | 369 | 7368263 | ||
| DQ198369.1 | - | 2 | 165 | |||

Btla
Genomic Map