siRNA Details
| Gene Symbol: Cfd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077642.1 | NP_001071110.1 | 5 | 737 | ||
| GenBank mRNA | M92059.1 | - | 5 | 734 | ||
| S73894.1 | - | 5 | 737 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfd| Gene Symbol: Cfd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077642.1 | NP_001071110.1 | 5 | 737 | ||
| GenBank mRNA | M92059.1 | - | 5 | 734 | ||
| S73894.1 | - | 5 | 737 | |||