siRNA Details
| Gene Symbol: Rgs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019344.2 | NP_062217.1 | 6 | 685 | ||
| XM_006250015.2 | XP_006250077.1 | 5 | 556 | |||
| XM_006250016.2 | XP_006250078.1 | 6 | 500 | |||
| XM_006250017.2 | XP_006250079.1 | 6 | 850 | |||
| XM_008769675.1 | XP_008767897.1 | 6 | 851 | |||
| XM_008769676.1 | XP_008767898.1 | 6 | 1016 | |||
| GenBank mRNA | AB006013.1 | - | 5 | 594 | ||
| BC089064.1 | - | 6 | 685 | 7317138 | ||
| U32432.1 | - | 1 | 22 | |||

Rgs8
Genomic Map