siRNA Details
| Gene Symbol: Rassf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019365.3 | NP_062238.1 | 6 | 1241 | ||
| XM_006249796.2 | XP_006249858.1 | 6 | 2102 | |||
| XM_008769494.1 | XP_008767716.1 | 5 | 920 | |||
| GenBank mRNA | AF002251.1 | - | 6 | 1241 | ||

Rassf5
Genomic Map