siRNA Details
| Gene Symbol: Psmb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031629.1 | NP_113817.1 | 2 | 253 | ||
| GenBank mRNA | L17127.1 | - | 2 | 253 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmb4| Gene Symbol: Psmb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031629.1 | NP_113817.1 | 2 | 253 | ||
| GenBank mRNA | L17127.1 | - | 2 | 253 | ||