siRNA Details
| Gene Symbol: Fkbp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022675.1 | NP_073166.1 | 4 | 244 | ||
| XM_006239860.2 | XP_006239922.1 | 3 | 383 | |||
| GenBank mRNA | D86642.1 | - | 4 | 244 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fkbp1b| Gene Symbol: Fkbp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022675.1 | NP_073166.1 | 4 | 244 | ||
| XM_006239860.2 | XP_006239922.1 | 3 | 383 | |||
| GenBank mRNA | D86642.1 | - | 4 | 244 | ||