siRNA Details
| Gene Symbol: Ramp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031646.1 | NP_113834.1 | 4 | 405 | ||
| GenBank mRNA | AB028934.1 | - | 2 | 119 | ||
| AB030943.1 | - | 4 | 414 | |||
| AB042888.1 | - | 4 | 405 | |||
| AF162778.1 | - | 2 | 159 | |||
| AF181551.1 | - | 4 | 408 | |||
| EF595744.1 | - | 4 | 329 | |||
| FQ226087.1 | - | 4 | 430 | |||

Ramp2
Genomic Map