siRNA Details
| Gene Symbol: Rgs19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021661.2 | NP_067693.1 | 5 | 645 | ||
| XM_006235801.2 | XP_006235863.1 | 5 | 3406 | |||
| XM_008762540.1 | XP_008760762.1 | 4 | 918 | |||
| XM_008762541.1 | XP_008760763.1 | 4 | 385 | |||
| XM_008762542.1 | XP_008760764.1 | 4 | 381 | |||
| GenBank mRNA | AF068136.1 | - | 4 | 353 | ||
| BC088141.1 | - | 5 | 645 | 7377022 | ||

Rgs19
Genomic Map