siRNA Details
| Gene Symbol: Nxph3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021679.1 | NP_067711.1 | 2 | 685 | ||
| GenBank mRNA | AF042713.1 | - | 2 | 685 | ||
| BC166603.1 | - | 607 | 7936112 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nxph3| Gene Symbol: Nxph3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021679.1 | NP_067711.1 | 2 | 685 | ||
| GenBank mRNA | AF042713.1 | - | 2 | 685 | ||
| BC166603.1 | - | 607 | 7936112 | |||