siRNA Details
| Gene Symbol: Ereg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021689.1 | NP_067721.1 | 4 | 417 | ||
| GenBank mRNA | AF074952.1 | - | 4 | 417 | ||
| DQ266371.1 | - | 2 | 145 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ereg| Gene Symbol: Ereg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021689.1 | NP_067721.1 | 4 | 417 | ||
| GenBank mRNA | AF074952.1 | - | 4 | 417 | ||
| DQ266371.1 | - | 2 | 145 | |||