siRNA Details
| Gene Symbol: Lin7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021851.1 | NP_068623.1 | 4 | 353 | ||
| GenBank mRNA | AF090136.1 | - | 4 | 353 | ||
| FQ212254.1 | - | 385 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lin7c| Gene Symbol: Lin7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021851.1 | NP_068623.1 | 4 | 353 | ||
| GenBank mRNA | AF090136.1 | - | 4 | 353 | ||
| FQ212254.1 | - | 385 | ||||