siRNA Details
| Gene Symbol: Capn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031673.2 | NP_113861.1 | 2 | 448 | ||
| GenBank mRNA | AB113362.1 | - | 2 | 355 | ||
| AB113363.1 | - | 2 | 355 | |||
| AF227909.3 | - | 2 | 348 | |||
| BC085725.1 | - | 2 | 448 | 7123652 | ||

Capn10
Genomic Map