siRNA Details
Gene Symbol: Capn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031673.2 | NP_113861.1 | 4 | 876 | ||
GenBank mRNA | AB113362.1 | - | 4 | 783 | ||
AB113363.1 | - | 4 | 783 | |||
AF227909.3 | - | 4 | 776 | |||
BC085725.1 | - | 4 | 876 | 7123652 |
