siRNA Details
| Gene Symbol: Otulin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729016.4 | 7 | 1144 | |||
| XM_006232088.2 | XP_006232150.1 | 7 | 1141 | |||
| GenBank mRNA | BC158613.1 | - | 7 | 1099 | 7384798 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otulin| Gene Symbol: Otulin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729016.4 | 7 | 1144 | |||
| XM_006232088.2 | XP_006232150.1 | 7 | 1141 | |||
| GenBank mRNA | BC158613.1 | - | 7 | 1099 | 7384798 | |