siRNA Details
| Gene Symbol: Prl8a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022537.1 | NP_071982.1 | 5 | 477 | ||
| XM_006253909.1 | XP_006253971.1 | 5 | 451 | |||
| GenBank mRNA | AB000107.1 | - | 5 | 477 | ||
| BC098917.1 | - | 5 | 457 | 7445772 | ||

Prl8a7
Genomic Map