siRNA Details
| Gene Symbol: Scamp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031726.1 | NP_113914.1 | 8 | 936 | ||
| GenBank mRNA | AF240784.1 | - | 8 | 936 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scamp5| Gene Symbol: Scamp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031726.1 | NP_113914.1 | 8 | 936 | ||
| GenBank mRNA | AF240784.1 | - | 8 | 936 | ||