siRNA Details
| Gene Symbol: Tas2r105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023999.1 | NP_076489.1 | 1 | 515 | ||
| GenBank mRNA | AF227146.1 | - | 1 | 515 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r105| Gene Symbol: Tas2r105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023999.1 | NP_076489.1 | 1 | 515 | ||
| GenBank mRNA | AF227146.1 | - | 1 | 515 | ||