siRNA Details
| Gene Symbol: Dnajc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024161.2 | NP_077075.1 | 5 | 705 | ||
| XM_008762544.1 | XP_008760766.1 | 5 | 754 | |||
| XM_008762545.1 | XP_008760767.1 | 6 | 795 | |||
| GenBank mRNA | AF323955.1 | - | 5 | 705 | ||
| FQ223796.1 | - | 5 | 746 | |||
| S81917.1 | - | 4 | 605 | |||
| U39320.1 | - | 5 | 705 | |||

Dnajc5
Genomic Map