siRNA Details
| Gene Symbol: Lat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030853.1 | NP_110480.1 | 5, 6 | 544 | ||
| GenBank mRNA | AJ001184.1 | - | 5, 6 | 544 | ||
| BC088130.1 | - | 5, 6 | 504 | 7382238 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lat| Gene Symbol: Lat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030853.1 | NP_110480.1 | 5, 6 | 544 | ||
| GenBank mRNA | AJ001184.1 | - | 5, 6 | 544 | ||
| BC088130.1 | - | 5, 6 | 504 | 7382238 | ||