siRNA Details
| Gene Symbol: Rala | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031093.2 | NP_112355.1 | 3 | 482 | ||
| XM_006253993.2 | XP_006254055.1 | 4 | 686 | |||
| XM_008771763.1 | XP_008769985.1 | 4 | 500 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rala| Gene Symbol: Rala | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031093.2 | NP_112355.1 | 3 | 482 | ||
| XM_006253993.2 | XP_006254055.1 | 4 | 686 | |||
| XM_008771763.1 | XP_008769985.1 | 4 | 500 | |||