siRNA Details
| Gene Symbol: Psmd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031331.1 | NP_112621.1 | 6 | 579 | ||
| XM_006232924.2 | XP_006232986.1 | 6 | 584 | |||
| GenBank mRNA | AB017188.1 | - | 6 | 579 | ||
| AF175575.1 | - | 6 | 520 | |||
| BC060559.1 | - | 6 | 529 | 6891370 | ||
| FQ209672.1 | - | 5 | 381 | |||

Psmd4
Genomic Map