siRNA Details
| Gene Symbol: Mchr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031758.1 | NP_113946.1 | 2 | 992 | ||
| GenBank mRNA | AF008650.1 | - | 2 | 992 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mchr1| Gene Symbol: Mchr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031758.1 | NP_113946.1 | 2 | 992 | ||
| GenBank mRNA | AF008650.1 | - | 2 | 992 | ||
Set Membership: |
|