siRNA Details
| Gene Symbol: Nfe2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031789.2 | NP_113977.1 | 5 | 1771 | ||
| XM_006234396.2 | XP_006234458.1 | 5 | 1940 | |||
| XM_006234397.2 | XP_006234459.1 | 5 | 1885 | |||
| XM_006234398.2 | XP_006234460.1 | 5 | 2492 | |||
| GenBank mRNA | AF037350.1 | - | 5 | 1773 | ||
| BC061724.1 | - | 5 | 1807 | 5598313 | ||
| FQ220760.1 | - | 1 | 608 | |||
| FQ221441.1 | - | 3 | 2716 | |||

Nfe2l2
Genomic Map