siRNA Details
| Gene Symbol: Scd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031841.1 | NP_114029.1 | 2 | 494 | ||
| GenBank mRNA | AB032243.1 | - | 2 | 494 | ||
| BC061737.1 | - | 2 | 436 | 5599134 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scd2| Gene Symbol: Scd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031841.1 | NP_114029.1 | 2 | 494 | ||
| GenBank mRNA | AB032243.1 | - | 2 | 494 | ||
| BC061737.1 | - | 2 | 436 | 5599134 | ||