siRNA Details
Gene Symbol: Cxxc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233339.2 | XP_006233401.1 | 3 | 957 | ||
XM_006233341.2 | XP_006233403.1 | 2 | 872 | |||
XM_008761529.1 | XP_008759751.1 | 2 | 1033 | |||
XM_008761530.1 | XP_008759752.1 | 2 | 1033 | |||
XM_008761531.1 | XP_008759753.1 | 2 | 1033 | |||
GenBank mRNA | AF272158.1 | - | 1 | 96 |
