siRNA Details
| Gene Symbol: Puf60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191880.1 | NP_001178809.1 | 11 | 1374 | ||
| XM_006241894.2 | XP_006241956.1 | 11 | 1510 | |||
| XM_006241895.2 | XP_006241957.1 | 10 | 1459 | |||
| XM_006241896.2 | XP_006241958.1 | 10 | 1456 | |||
| XM_006241897.2 | XP_006241959.1 | 10 | 1408 | |||
| XM_006241898.2 | XP_006241960.1 | 9 | 1357 | |||
| XM_006241899.2 | XP_006241961.1 | 11 | 1803 | |||
| GenBank mRNA | AF165892.1 | - | 10 | 1363 | ||

Puf60
Genomic Map