siRNA Details
| Gene Symbol: Pgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053595.2 | NP_446047.1 | 3 | 588 | ||
| GenBank mRNA | BC087006.1 | - | 3 | 588 | 7193034 | |
| L40030.1 | - | 3 | 323 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgf| Gene Symbol: Pgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053595.2 | NP_446047.1 | 3 | 588 | ||
| GenBank mRNA | BC087006.1 | - | 3 | 588 | 7193034 | |
| L40030.1 | - | 3 | 323 | |||