siRNA Details
| Gene Symbol: Olr1867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101022.1 | NP_001094492.1 | 1 | 1029 | ||
| GenBank mRNA | D12820.1 | - | 1 | 1029 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1867| Gene Symbol: Olr1867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101022.1 | NP_001094492.1 | 1 | 1029 | ||
| GenBank mRNA | D12820.1 | - | 1 | 1029 | ||