siRNA Details
| Gene Symbol: LOC100125364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103356.2 | NP_001096826.1 | 2 | 665 | ||
| XM_006227969.2 | XP_006228031.1 | 2 | 598 | |||
| XM_006227970.2 | XP_006228032.1 | 2 | 576 | |||
| XM_008758746.1 | XP_008756968.1 | 2 | 671 | |||
| GenBank mRNA | BC079175.1 | - | 2 | 665 | 7130104 | |

LOC100125364
Genomic Map