siRNA Details
| Gene Symbol: Pnrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103360.1 | NP_001096830.1 | 3 | 698 | ||
| GenBank mRNA | BC081903.1 | - | 3 | 698 | 7110880 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnrc2| Gene Symbol: Pnrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103360.1 | NP_001096830.1 | 3 | 698 | ||
| GenBank mRNA | BC081903.1 | - | 3 | 698 | 7110880 | |