siRNA Details
| Gene Symbol: Nudcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103362.1 | NP_001096832.1 | 727 | |||
| GenBank mRNA | BC107925.1 | - | 727 | 7443892 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nudcd3| Gene Symbol: Nudcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103362.1 | NP_001096832.1 | 727 | |||
| GenBank mRNA | BC107925.1 | - | 727 | 7443892 | ||