siRNA Details
| Gene Symbol: Mroh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103364.1 | NP_001096834.1 | 4 | 324 | ||
| GenBank mRNA | BC093388.1 | - | 4 | 324 | 7389659 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mroh1| Gene Symbol: Mroh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103364.1 | NP_001096834.1 | 4 | 324 | ||
| GenBank mRNA | BC093388.1 | - | 4 | 324 | 7389659 | |