siRNA Details
| Gene Symbol: Zfp786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127594.1 | NP_001121066.1 | 4 | 1759 | ||
| XM_006236413.2 | XP_006236475.1 | 4 | 1816 | |||
| XM_006236414.2 | XP_006236476.1 | 4 | 1902 | |||
| GenBank mRNA | BC166479.1 | - | 4 | 1759 | 9102251 | |

Zfp786
Genomic Map