siRNA Details
| Gene Symbol: LOC100158225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127606.1 | NP_001121078.1 | 7 | 818 | ||
| GenBank mRNA | BC166509.1 | - | 7 | 818 | 7127250 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100158225| Gene Symbol: LOC100158225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127606.1 | NP_001121078.1 | 7 | 818 | ||
| GenBank mRNA | BC166509.1 | - | 7 | 818 | 7127250 | |