siRNA Details
| Gene Symbol: Sncb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080777.2 | NP_542955.2 | 2, 3 | 329 | ||
| XM_006253594.2 | XP_006253656.1 | 3, 4 | 530 | |||
| GenBank mRNA | BC087579.1 | - | 2, 3 | 329 | 7314241 | |
| D17764.1 | - | 2, 3 | 185 | |||

Sncb
Genomic Map