siRNA Details
| Gene Symbol: Sncb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080777.2 | NP_542955.2 | 5, 6 | 575 | ||
| XM_006253594.2 | XP_006253656.1 | 6, 7 | 776 | |||
| GenBank mRNA | BC087579.1 | - | 5, 6 | 575 | 7314241 | |
| D17764.1 | - | 5, 6 | 431 | |||

Sncb
Genomic Map