siRNA Details
| Gene Symbol: Prnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102431.1 | NP_001095901.1 | 1 | 284 | ||
| XM_006235056.2 | XP_006235118.1 | 2 | 440 | |||
| XM_006235057.2 | XP_006235119.1 | 2 | 440 | |||
| XM_006235058.2 | XP_006235120.1 | 2 | 394 | |||
| GenBank mRNA | BC079436.1 | - | 1 | 296 | ||
| GQ245669.1 | - | 1 | 284 | |||

Prnd
Genomic Map